Learn More
STEMCELL Technologies EasySep™ Direct Human PBMC Isolation Kit RoboSep™
Product Code. 17137941
Encompass
Change view
Click to view available options
Quantity:
100 mL Whole Blood
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17137941 100 mL Whole Blood 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17137941 Supplier STEMCELL Technologies Supplier No. 19654RF
EasySep™ kit with RoboSep™ buffer and RoboSep™ filter tips
  • The EasySep™ Direct Human PBMC Isolation Kit is designed to isolate highly purified peripheral blood mononuclear cells (PBMCs) from fresh whole blood, buffy coat, bone marrow, cord blood, or leukapheresis products by negative selection
  • Red blood cells (RBCs), platelets, and unwanted cells are targeted for removal with antibody complexes and magnetic particles and separated using an EasySep™ magnet
  • Untouched PBMCs are simply collected into a new tube and are immediately available for downstream applications such as flow cytometry, culture, or DNA/RNA extraction
Specifications
Cell Type B Cells, Lymphocytes, Monocytes, Mononuclear Cells, NK Cells, T Cells
Quantity 100 mL Whole Blood