missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies EasySep™ Human Extracellular Vesicle (CD81) Positive Selection Kit

Product Code. 17127811
Encompass
Click to view available options
Quantity:
20 mL of biofluid
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 17127811

Brand: STEMCELL Technologies 17892

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Fast & easy isolation of human extracellular vesicles using immunomagnetic positive selection

  • The EasySep™ Human Extracellular Vesicle (CD81) Positive Selection Kit isolates highly purified human extracellular vesicles from plasma/serum and cell culture conditioned medium by immunomagnetic positive selection
  • Extracellular vesicles are labeled with Tetrameric Antibody Complexes recognizing CD81 and magnetic particles
  • Labeled extracellular vesicles are separated without columns using an EasySep™ magnet
  • Unwanted biofluid components are simply poured off, while desired extracellular vesicles remain in the tube
  • Isolated extracellular vesicles are available for downstream applications such as DNA/RNA extraction, Western blot, or mass spectometry​

Specifications

Cell Type Other
For Use With (Application) Cell Isolation
Quantity 20 mL of biofluid
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.