missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies EasySep™ Human Pan-Extracellular Vesicle Positive Selection Kit
Click to view available options
:
20 mL of biofluid
Description
- The EasySep™ Human Pan-Extracellular Vesicle Positive Selection Kit isolates highly purified human extracellular vesicles from plasma/serum and cell culture conditioned medium by immunomagnetic positive selection
- Extracellular vesicles are labeled with Tetrameric Antibody Complexes recognizing CD9, CD81, and CD63, and magnetic particles
- Labeled extracellular vesicles are separated without columns using an EasySep™ magnet
- Unwanted biofluid components are simply poured off, while desired extracellular vesicles remain in the tube
- Isolated extracellular vesicles are available for downstream applications such as DNA/RNA extraction, Western blot, or mass spectometry
Specifications
Specifications
| Cell Type | Other |
| For Use With (Application) | Cell Isolation |
| Quantity | 20 mL of biofluid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction