Learn More
STEMCELL Technologies EasySep™ Mouse B Cell Isolation Kit
Description
- The EasySep™ Mouse B Cell Isolation Kit is designed to isolate B cells from single-cell suspensions of splenocytes or other tissues by negative selection
- Unwanted cells are targeted for removal with biotinylated antibodies directed against non-B cells and streptavidin-coated magnetic particles (RapidSpheres™ )
- Labeled cells are separated using and EasySep™ magnet without the use of columns
- Desired cells are poured off into a new tube
- This product replaces the EasySep™ Mouse B Cell Enrichment Kit (Catalog #19754) for even faster cell isolations
Specifications
Specifications
| Cell Type | B Cells |
| For Use With (Application) | Cell Isolation |
| Quantity | 1 x 109 Cells |