Learn More
STEMCELL Technologies EasySep™ Mouse T Cell Isolation Kit
Product Code. 16617665
Encompass
Change view
Click to view available options
Quantity:
1 x 109 Cells
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16617665 1 x 109 Cells Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16617665 Supplier STEMCELL Technologies Supplier No. 19851
Immunomagnetic isolation of untouched mouse T cells
  • The EasySep™ Mouse T Cell Isolation Kit is designed to isolate T cells from single-cell suspensions of splenocytes or other tissues by negative selection
  • Unwanted cells are targeted for removal with biotinylated antibodies directed against non-T cells and streptavidin-coated magnetic particles
  • Labeled cells are separated using an EasySep™ magnet without the use of columns
  • Desired cells are poured off into a new tube
  • This product replaces the EasySep™ Mouse T Cell Enrichment Kit (Catalog #19751) for even faster cell isolations
Specifications
Cell Type T Cells
For Use With (Application) Cell Isolation
Quantity 1 x 109 Cells