Learn More
STEMCELL Technologies EasySep™ Release Human CD3 Positive Selection Kit
Click to view available options
Quantity:
1 x 109 cells
Unit Size:
1 set
Description
- Isolate highly purified and magnetic particle-free CD3+ cells from peripheral blood mononuclear cells or washed leukapheresis samples in as little as 30 minutes
- Using immunomagnetic positive selection, CD3+ cells are labeled with antibodies and magnetic particles, then separated without columns using an EasySep magnet
- Unwanted cells are poured off, while desired cells remain in the tube
- Bound magnetic particles are then removed from the isolated CD3+ cells, which are immediately available for downstream use
- For even higher efficiency, the process can be automated using RoboSep
- This kit is designed for cell therapy research applications, but may be qualified for use as an ancillary material following the framework outlined in USP
- STEMCELL Technologies can work with you to qualify this reagent as an ancillary material under an approved investigational new drug (IND) or clinical trial application (CTA)
Specifications
Specifications
| Cell Type | T Cells |
| Quantity | 1 x 109 cells |