Learn More
Ebro Electronic™ EBI 25-TE Temperature Logger
Product Code. 11763529
Click to view available options
Temperature Range (Metric):
-40°C to +85°C
Unit Size:
Each
Product Code. 11763529
Brand: Ebro Electronic™ 13406201A
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Temperature logger with external probe.
Probe PT 100 for TFX 430, PT 100, 200 mm, blunt
TRUSTED_SUSTAINABILITY
Specifications
Type Datalogger
Display Type LCD
Data Memory 288 Readings
Calibration Calibrated
Alarm Types E-mail
Battery Type Lithium
Battery Life <2 Years
Channels 1
Certifications/Compliance Calibration certificate
IP Rating IP67
Model EBI 25-TE
Sensor Types NTC
For Use With (Application) Logging and recording of temperatures, Storage monitoring, Deep-freezers, Refrigerators and refrigerated display cases
Accuracy (Temperature) ±0,5 at –20 to 40, ±0,8 to–30 to –20 /40 to 60, ±1,5 at –40 to –30 / 60 to 85 °C
Temperature Resolution 0.1°C
Temperature Range (Metric) -40°C to +85°C
Dimensions (L x W x H) 95 x 48 x 27 mm
Length (Metric) 95 mm
Weight (Metric) 65 g