Learn More
Elma™ EC 60 Cleaner
Product Code. 10300422
Change view
Click to view available options
Volume (Metric):
1 L
25 L
Unit Size:
1 L
25 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
10300422 25 L 25 L
10407041 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10300422 Supplier Elma™ Supplier No. 5810010000
Ideal cleaning concentrate for ultrasonic and immersion cleaning.
  • Suitable for acidic thorough cleaning of medical instruments, implants prostheses, workpieces made of stainless-steel, metal, ceramic, glass and plastics.
  • Also suitable for removing of acid-soluble cements and for laboratory equipment.
  • Not suitable for light metals.
  • Removes metal oxides and minderal deposits like lime, tarnished spots and other deposits, mineral oils and fats/greases.
Specifications
Product Type EC 60 Cleaner
Form Liquid
pH 0 to 3
Appearance Yellow
Boiling Point >100°C
Certifications/Compliance Annex VII A, EC 648/2004
Composition Non-ionic surfactants <5%, Phosphoric Acid >30%
Concentration Ultrasonic bath 1 to 10% at 40 to 70°C, in immersion bath >3%
Content And Storage Above -5°C
For Use With (Application) For cleaning of medical and dental instruments
Volume (Metric) 25 L
Show More Show Less
compliance-icons
Product Identifier
  • elma clean 60
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308-IF exposed or concerned:.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: phosphoric acid, isotridecanol, ethoxylated