missing translation for 'onlineSavingsMsg'
Learn More

Elma™ EC 60 Cleaner

Product Code. 10407041
Change view
Click to view available options
Volume (Metric):
1 L
25 L
Unit Size:
1 L
25 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
10407041 1 L 1 L
10300422 25 L 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10407041 Supplier Elma™ Supplier No. 5809900000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ideal cleaning concentrate for ultrasonic and immersion cleaning.

  • Suitable for acidic thorough cleaning of medical instruments, implants prostheses, workpieces made of stainless-steel, metal, ceramic, glass and plastics.
  • Also suitable for removing of acid-soluble cements and for laboratory equipment.
  • Not suitable for light metals.
  • Removes metal oxides and minderal deposits like lime, tarnished spots and other deposits, mineral oils and fats/greases.

Specifications

Product Type EC 60 Cleaner
Form Liquid
pH 0 to 1
Appearance Yellow
Boiling Point >100°C
Certifications/Compliance Annex VII A, EC 648/2004
Composition Non-ionic surfactants <5%, Phosphoric Acid >30%
Concentration Ultrasonic bath 1 to 10% at 40 to 70°C, in immersion bath >3%
Container Type Bottle
Content And Storage Above -5°C
For Use With (Application) For cleaning of medical and dental instruments
Volume (Metric) 1 L
Show More Show Less
compliance-icons
Product Identifier
  • EC 60
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.