missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies EC23
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
Description
- EC23 is a retinoic acid receptor (RAR) agonist with pan-RAR activity (EC50: RARα 41 nM, RARβ 0.5 nM, RARγ 0.4 nM), while having no appreciable activity for retinoid X receptors (RXR; EC50> 10μM for all; Gambone et al.) It is a photostable synthetic analog of all-trans retinoic acid (ATRA; Christie et al. 2008)
- EC23 also weakly activates aryl hydrocarbon receptor (Gambone et al.)
- Induces neural differentiation of human pluripotent stem cells, similarly to ATRA (Christie et al. 2010; Clemens et al.)
- Induces neuronal differentiation of human fetal neural progenitor cell line ReNcell 197VM (Christie et al. 2010)
Specifications
Specifications
| CAS | 104561-41-3 |
| Synonym | AGN 190205; BASF 46928 |
| Quantity | 10 mg |
| Molecular Formula | C23H24O2 |
| Purity | ≥98% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction