missing translation for 'onlineSavingsMsg'
Learn More

Kolmi Hopen™ Eco Isolation Gowns with Back Laces

Product Code. 17760159 Shop All Medicom Products
Change view
Click to view available options
Color:
Green
Sky Blue
White
Yellow
Unit Size:
Pack of 10
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
17760159 Green Pack of 10
17770159 White Pack of 10
17780159 Sky Blue Pack of 10
17790159 Yellow Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 17760159 Supplier Kolmi Hopen™ Supplier No. J40451

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

These isolation gowns are complete with elasticated wrists and easy tie closures for busy healthcare workers.

  • Light and breathable
  • Collarless model with elasticated cuffs
  • Dorsal closure with laces at waistband and collar
  • Worn by healthcare workers and visitors over clothing
  • Protects patients from risk of external infection

Specifications

Size Universal
For Use With (Application) Dental, Medical
Gender Unisex
No. of Pockets None
Product Type Isolation Gown
Material Polypropylene
Color Green
Closure Type Back Ties
Disposable Single-use
Sterility Non-sterile
Weight (Metric) 17 g/m2
Autoclavable Not Autoclavable
No. per Pack 10 Pk.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.