Learn More
Skytec™ Eco Rhodium Gloves
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Unit Size:
10 pairs
Beskrivning
- PU-Coated Glove with High Cut Resistance
- Steel and glass fiber free to avoid any risks of irritation
- Durable PU palm coating ensures good protection against abrasion and secure grip in dry and greasy conditions
- Reinforced thumb crotch enhances durability
- Touchscreen capabilities to allow usage with screen devices
- Seamless knit prevents irritation
- RCS (Recycled Claim Standard) certified
- Elasticated wrist provides a secure fit
- 1 pair = 1 recycled 500 mL (PET) plastic bottle
- 18 g of CO2 reduced vs virgin polyester alternative
Specifikationer
Specifikationer
| Certifications/Compliance (Europe) | CE II, UKCA, ANSI/ISEA 105-2016: Cut A5, EN 388:2016 + A1:2018 4X43E, EN 21420:2020 |
| Coating Material | Polyurethane |
| Color | Gray, Black |
| Cut Resistance Rating | Level E |
| Hand | Ambidextrous |
| Latex Free | Yes |
| Material | Polyester |
| Product Type | Cut-resistant Gloves |
| Size | Large |
| EN388 Standard | 4X43E |
| Visa mer |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering