Description
- Made of a seamless liner, palm dipped into micro-foam nitrile coating.
- The micro-foam nitrile coating provides excellent tactile sensitivity and dexterity vs. foam nitrile coating.
- The micro-foam nitrile coating provides a good grip and excellent abrasion resistance, compatible with slight oil.
- Very comfortable to wear, the breathable coating keeps hands cool and dry.
Specifications
Specifications
| Coating Material | Nitrile Foam |
| Color | Black |
| Cut Resistance Rating | Level B |
| Hand | Specific |
| Latex Free | Yes |
| Material | HPPE, Nylon, Polyester, Spandex, Foam Nitrile |
| Product Type | Industrial Gloves |
| Size | 11 |