missing translation for 'onlineSavingsMsg'
Learn More

Ansell EDGE™ 48-703 Seamless Knit Polyethylene Gloves with Leather Palm Pad

Product Code. 30259524 Shop All Ansell Products
Click to view available options
Size:
9
Unit Size:
12 pairs
This item is not returnable. View return policy

Product Code. 30259524

Brand: Ansell 48703090

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Gloves combine cut, abrasion, and thermal resistance with a leather palm pad for additional cushion and support during metal stamping applications.

  • Seamless construction with a knit wrist provides a close fit for a high level of dexterity
  • Re-inforcement protects the user when handling hot or sharp objects
  • Enhanced grip capabilities in dry or slightly oily environments
  • Leather palm pad offers additional cushioning and protection
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance (Europe) EN 420:2003 + A1:2009; Category III
Construction Seamless Knit
Coating Material Nitrile
Color Gray
Exterior Finish No Texture
Hand Specific
Latex Free No
Material Polyethylene, Fiberglass
Product Type Seamless Knit Gloves
Size 9
Style Clute
EN388 Standard 4X43D
For Use With (Application) Automotive Assembly, Machinery/Equipment Operation, Metal Fabrication, and Oil/Gas Industries
Glove Cuff Style Knit
Palm Type Leather
Series EDGE™ 48-703
Yarn Gauge 13
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.