missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EDNRA/Endothelin R Type A Antibody (485709), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB65381Z
This item is not returnable.
View return policy
Description
EDNRA/Endothelin R Type A Monoclonal antibody specifically detects EDNRA/Endothelin R Type A in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
EDNRA/Endothelin R Type A | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
485709 | |
Flow Cytometry, CyTOF-ready | |
endothelin receptor type A, endothelin-1 receptor, endothelin-1-specific receptor, ET1-specific type, ETAR, ETRA, G protein-coupled receptor, hET-AR | |
NS0 mouse myeloma cell line transfected with human EDNRA/Endothelin R Type A, Accession # NP_001948 | |
0.1 mL | |
GPCR, Neuroscience, Signal Transduction | |
1909 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction