Learn More
Invitrogen™ EGFR (ErbB1) [L861Q] Recombinant Human Protein
The EGFR L861Q mutation is an Iressa-responsive mutant observed in the clinic.
Brand: Invitrogen™ PV4191
Additional Details : Weight : 0.00900kg
Description
The epidermal growth factor receptor is a receptor tyrosine kinase that signals in response to various growth factors. Overexpression has been linked to numerous types of cancer and EGFR is the target of both biological and small molecular therapeutics.
Specifications
NP_005219.2; L861Q | |
50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
1mg | |
RUO - research use only | |
EGFR (ErbB1) L861Q | |
Human | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
90.5kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Cytoplasmic | |
EGF Receptor (ErbB) Kinase Family | |
Recombinant | |
Baculovirus |