missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EGFR (ErbB1) [T790M] Recombinant Human Protein
The epidermal growth factor receptor is a receptor tyrosine kinases that signals in response to various growth factors.
1826.00€ - 7890.00€
Specifications
Accession Number | NP_005219.2; T790M |
---|---|
Formulation | 50mM Tris pH 7.5, 150mM NaCl, 0.02% Trition X-100, 0.5mM EDTA, 2mM DTT, 50% Glycerol |
Molecular Weight (g/mol) | 90.5kDa |
Storage Requirements | Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Regulatory Status | RUO - research use only |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15498643
|
Invitrogen™
PR8052A |
100μg |
1826.00€
100µg |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15408763
|
Invitrogen™
PV4804 |
1mg |
7890.00€
1mg |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Overexpression of EGFR has been linked to numerous types of cancer and EGFR is the target of both biological and small molecular therapeutics. The T790M mutant of EGFR has arisen in the clinics and displays resistance to Erlotinib (Tarceva) and Gefitinib (Iressa).
Specifications
NP_005219.2; T790M | |
90.5kDa | |
RUO - research use only | |
EGFR (ErbB1) T790M | |
Human | |
GST Tag | |
Frozen |
50mM Tris pH 7.5, 150mM NaCl, 0.02% Trition X-100, 0.5mM EDTA, 2mM DTT, 50% Glycerol | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Cytoplasmic | |
EGF Receptor (ErbB) Kinase Family | |
Recombinant | |
Baculovirus |