Learn More
Kolmi Hopen™ Elasticated SMS Gown with Collar and Zipper
GREENER_CHOICE
Product Code. p-7227155 Shop All Medicom Products
Click to view available options
Size:
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 50
Product Code. 17740119
Brand: Kolmi Hopen™ 40.507/03
Only null left
Add to basket
Add to basket
The polyethylene film of these gowns makes it possible to reduce the particulate release of fibers in the industrial workplace.
  • Lightweight polypropylene is lined with a polyethylene film for better hydrophobic sealing
  • With collar and elasticated cuffs
  • Suitable for protected environments
  • Individual packaging for controlled hygiene and distribution
Specifications
Size Large
Material SMS, Polyethylene
Color White
Closure Type Zipper
Disposable Single-use
Certifications/Compliance (Europe) ISO 9001, EN 1149-5
Weight (Metric) 65 g/m2
No. per Pack 50 Pk.