Learn More
SI Analytics™ Electrolyte Bridge
SI Analytics™ Electrolyte Bridge is constructed of glass and designed for electrode installation.
Brand:  SI Analytics™ 285104209
Additional Details : Weight : 0.80000kg
Product Code. 10589201
  • 146.00€ / Each
Estimated Shipment: 31-07-2024
to see stock.
Add to basket
Description
Description
  • Standard taper NS 14.5
  • Sleeve NS 14.5
  • For electrode installation
Specifications
Specifications
Ceramic
Electrolyte Bridge
B 511
103 mm