Learn More
Electrolytes and Cleaning Solutions, WTW™
Click to view available options
Quantity:
50 mL
Unit Size:
Each
Description
- Solutions for operation, cleaning and storage of electrodes
- Simple refilling and safe storage warranted
- Used as refill solutions for liquid filled electrodes as well as a storage solution for all types of pH electrodes
- With the pepsin cleaning solution, you can remove protein precipitation from the diaphragm of electrodes with liquid electrolyte
Specifications
Specifications
| Density | 1.13g/cm3 |
| Concentration | 3 mol/L |
| For Use With (Application) | Operation, cleaning and storage of electrodes |
| pH | 7 |
| Physical Form | Liquid |
| Quantity | 50 mL |
| Packaging | Bottle |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur