missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ ELECTROSOFT COMPOSITE Class 0 Latex Gloves

Honeywell™ ELECTROSOFT COMPOSITE Class 0 Latex Gloves for electrical work are designed to be used in Arc Flash environments.

Brand:  Honeywell™ 2092002-10

Additional Details : Weight : 0.70000kg

 View more versions of this product

Product Code. 12657603

  • 83.68€ / Each


Estimated Shipment
08-10-2021
Add to basket

Description

Description

  • Insulating glove for electrical work
  • External layer in orange polychloroprene
  • Chlorinated inside
  • Category RC
  • ATPV 21.6/cal
  • Dielectric natural latex composition on chlorinated interior base makes it very comfortable
  • Product appreciated for its long life and precision
  • Patterned finish gives it a highly appreciated grip in damp conditions
  • The outer orange elastomer layer provides excellent mechanical resistance
  • Its anatomical shape makes flexing easier while ensuring a high level of protection
  • Specificaly designed to be used in Arc Flash environement
Specifications

Specifications

Orange
36 cm
Latex and polychloroprene
10
EN 60903-2003, CEI 903-2002, IEC 61482-1-2
1.1 mm
Videos
SDS
Documents