Learn More
GVS Elipse Low Profile Mask with A2P3 Filters
Click to view available options
:
GVS Elipse Low Profile Mask
Description
Elipse low profile mask with A2P3 filters for organic gases and vapors until 5000 ppm and dust.
Specifications
Specifications
Color | Blue |
Disposable | Reusable |