Learn More
GVS Elipse Low Profile Mask with A2P3 Filters
GREENER_CHOICE
Product Code. 18479645
Click to view available options
:
GVS Elipse Low Profile Mask
Elipse low profile mask with A2P3 filters for organic gases, vapors, and dust.
Elipse low profile mask with A2P3 filters for organic gases and vapors until 5000 ppm and dust.
Specifications
Color Blue
Disposable Reusable