Learn More
Invitrogen™ Silencer™ Select MALAT1 Positive Control siRNA
Silencer™ Select MALAT1 Positive Control siRNA is extensively validated and an ideal control for many aspects of an siRNA experiment. 5nmol is provided.
Brand: Invitrogen™ 4455877
Additional Details : Weight : 0.01000kg
Description
The Silencer™ MALAT1 positive control is extensively validated and an ideal “test” siRNA for those just beginning siRNA experiments, because it is validated to work in multiple cell lines. Because it targets MALAT1 mRNA, many assays are available to measure its effects, including real-time RT-PCR.What are Silencer™ Select siRNAs?Silencer™ Select siRNAs incorporate the latest improvements in siRNA design, off-target effect prediction algorithms, and chemical modifications, to yield siRNAs with unrivalled efficacy, potency, and specificity. The result is fewer failed experiments due to poor silencing, and cleaner, more consistent phenotypic data.- Premium-quality siRNA, highly purified and ready to use
- Functionally tested in several common cell lines
- Include Silencer™ Select modifications for enhanced specificity
- For use in human, mouse, and rat cells
Specifications
The siRNA is provided dried down in a single tube, along with Nuclease-free Water for resuspension, and should be stored at –20°C. | |
siRNA | |
Unconjugated | |
siRNA | |
5 nmol | |
MALAT1 |
Positive Control | |
Transfection, RNAi | |
Silencer™ Select, Ambion™ | |
HPLC | |
Dry Ice |
For Research Use Only. Not for use in diagnostic procedures.