Learn More
Merck Durapore™ Membrane Filters, 0.22 μm
Recommended when maximum recovery is important. Durapore™ PVDF Membranes: 0.22μ Pore Size provide high flow rates and throughput, low extractables, broad chemical compatibility and low protein binding. Available in diameters ranging from 13mm to 142mm.
148.00€ - 1115.00€
Specifications
Autoclavable | Not Autoclavable |
---|---|
Color | White |
Filter Type | Screen Filter |
Material | PVDF |
Pore Size | 0.22 μm |
Product Code | Brand | Diameter (Metric) | Dimensions (L x W) | Wettability | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Dimensions (L x W) | Wettability | Price | Quantity & Availability | |||
10752901
|
Merck
GVWP04700 |
47 mm | Hydrophilic |
367.00€
Pack of 100 |
In Stock
Log in to see stock. |
||||
10742901
|
Merck
GVWP02500 |
25 mm | Hydrophilic |
266.00€
Pack of 100 |
Estimated Shipment: 29-07-2025
Log in to see stock. |
||||
10074500
|
Merck
GVWP01300 |
13 mm | Hydrophilic |
179.00€
Pack of 100 |
Estimated Shipment: 24-07-2025
Log in to see stock. |
||||
10732731
|
Merck
GVWP14250 |
142 mm | Hydrophilic |
704.00€
Pack of 50 |
Estimated Shipment: 22-07-2025
Log in to see stock. |
||||
10146710
|
Merck
GVWP09050 |
90 mm | Hydrophilic |
402.00€
Pack of 50 |
Estimated Shipment: 01-08-2025
Log in to see stock. |
||||
10656741
|
Merck
GVHP04700 |
47 mm | Hydrophobic |
421.00€
Pack of 100 |
Estimated Shipment: 24-07-2025
Log in to see stock. |
||||
10646741
|
Merck
GVHP02500 |
25 mm | Hydrophobic |
300.00€
Pack of 100 |
Estimated Shipment: 09-09-2025
Log in to see stock. |
||||
10646551
|
Merck
GVHP01300 |
13 mm | Hydrophobic |
241.00€
Pack of 100 |
Estimated Shipment: 09-09-2025
Log in to see stock. |
||||
10696161
|
Merck
GVHP09050 |
90 mm | Hydrophobic |
565.00€
Pack of 50 |
Estimated Shipment: 09-09-2025
Log in to see stock. |
||||
10742151
|
Merck
GVHP14250 |
142 mm | Hydrophobic |
879.00€
Pack of 50 |
Estimated Shipment: 09-09-2025
Log in to see stock. |
||||
10479830
|
Merck
GVWP10050 |
100 mm | Hydrophilic |
549.00€
Pack of 50 |
Estimated Shipment: 05-08-2025
Log in to see stock. |
||||
15858077
|
Merck
GVWP07625 |
76 mm | Hydrophilic |
180.00€
Pack of 25 |
Estimated Shipment: 06-08-2025
Log in to see stock. |
||||
16339646
|
Merck
GVWP06225 |
63.5 mm | Hydrophilic |
148.00€
Pack of 25 |
Estimated Shipment: 09-09-2025
Log in to see stock. |
||||
10606171
|
Merck
GVWP00010 |
304 mm | 3.04 m x 304 mm | Hydrophilic |
1115.00€
Each |
Estimated Shipment: 09-09-2025
Log in to see stock. |
|||
10194073
|
Merck
GVHP00010 |
304 mm | 3.04 m x 304 mm | Hydrophobic | N/A | ||||
Description
Recommended when maximum recovery is important. Durapore™ PVDF Membranes: 0.22μ Pore Size provide high flow rates and throughput, low extractables, broad chemical compatibility and low protein binding. Available in diameters ranging from 13mm to 142mm.
Durapore™ membranes provide high flow rates and throughput, low extractables and broad chemical compatibility. Hydrophlic Durapore™ membrane binds far less protein than nylon, nitrocellulose, or PTFE membranes.- Features & Benefits
Specifications
Not Autoclavable | |
Screen Filter | |
0.22 μm | |
Filter Membranes |
White | |
PVDF | |
Non-sterile |