Learn More
Merck Millipore EZ-Fit™ Manifolds
General microbiological analysis
Brand:  Merck Millipore EZFITL0W01
Additional Details : Weight : 0.96000kg
 View more versions of this product
Product Code. 14812021
  • 1705.00€ / Each
Estimated Shipment date
from Supplier 13-07-2020
Add to basket
Description
Description
EZ-Fit™ Manifold is available with the new Microfil™ head for use with presterilized, ready-to-use funnels and membranes.
Specifications
Specifications
Yes
Microfil V or Microfil S filtration units
1-place