Learn More
Merck Millipore EZ-Fit™ Manifolds
General microbiological analysis
Brand:  Merck Millipore EZFITMIC06
Additional Details : Weight : 5.80000kg
 View more versions of this product
Product Code. 14882011
  • 4740.00€ / Each
Estimated Shipment date
from Supplier 19-06-2020
Add to basket
Description
Description
EZ-Fit™ Manifold is available with the new Microfil™ head for use with presterilized, ready-to-use funnels and membranes.
Specifications
Specifications
Yes
Microfil funnels and membranes
6-place