Learn More
Merck Accessories for Fluid Sampling Kit
Accessories for filter holders for sample preparation.
Brand:  Merck XX6403735
473.72 EUR valid until 2024-12-31
Use promo code "23269" to get your promotional price.
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10365521
  • 510.00€ / Each
Estimated Shipment: 22-05-2024
to see stock.
Add to basket
Description
Description
NPTF quick-release valve
Specifications
Specifications
Stainless Steel
Series 300 Stainless Steel
Laboratory Filter Holders or Cyclones
0.3 cm
Non-sterile
Hose Connector, 1/8 in. NPTF quick-release valve
1/8 in.