missing translation for 'onlineSavingsMsg'
Learn More

Merck Millipore Hydrophobic 3mm Edge Membranes

For microbiological and sterility testing of antibiotics and bacteriostatic drugs, as well as for filling machine operations

Brand:  Merck Millipore HAEP047A0

Additional Details : Weight : 0.01000kg Transport : UN number : 3270 Chem class : 4.1 Pack group : II


 View more versions of this product

Product Code. 12831566

  • 237.00€ / Pack of 100


Estimated Shipment: 30-07-2021
Add to basket

Description

Description

  • Nonwetting edge prevents solution intrusion under holder sealing rings
  • Hydrophobic edge membranes are standard MF-Type (mixed cellulose ester) filters
  • 47mm diameter

Specifications

Specifications

White
4.7 cm
Hydrophobic Edge
Plain
Mixed Cellulose Ester
Non-sterile
1.85 in.
Microbiological and sterility testing of antibiotics and bacteriostatic drugs, as well as for filling machine operations
Autoclave-packed (10 resealable envelopes containing 10 filters and 10 pads, requires autoclaving)
Membrane Filter
0.45 μm
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 0.45µm 25mm white gridded membrane filters made of mixed cellulose esters

Signal Word
  • Danger

Hazard Category
  • Flammable solid Category 1

Hazard Statement
  • H228-Flammable solid.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P370+P378-In case of fire: Use powder to extinguish.

Supplemental information
  • MIXTURE LIST-Contains : nitrocellulose, octylphenol polyethoxyethanol

SDS
Documents
Certificates