missing translation for 'onlineSavingsMsg'
Learn More

Merck Millipore Hydrophobic 3mm Edge Membranes

For microbiological and sterility testing of antibiotics and bacteriostatic drugs, as well as for filling machine operations

Brand:  Merck Millipore GSEP047S0

Additional Details : Weight : 0.02000kg Transport : UN number : 3270 Chem class : 4.1 Pack group : II


 View more versions of this product

Product Code. 12801566

  • 252.00€ / Pack of 100


Estimated Shipment: 27-07-2021
Add to basket

Description

Description

  • Nonwetting edge prevents solution intrusion under holder sealing rings
  • Hydrophobic edge membranes are standard MF-Type (mixed cellulose ester) filters
  • 47mm diameter

Specifications

Specifications

White
4.7 cm
Hydrophobic Edge
Plain
Mixed Cellulose Ester
Sterile
1.85 in.
Microbiological and sterility testing of antibiotics and bacteriostatic drugs, as well as for filling machine operations
Sterile-packed (10 resealable envelopes containing 10 filters and 10 pads, sterile)
Membrane Filter
0.22 μm
Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Flammable solid Category 1

Hazard Statement
  • H228-Flammable solid.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P370+P378-In case of fire: Use powder to extinguish.

Supplemental information
  • MIXTURE LIST-Contains : nitrocellulose

SDS
Documents
Certificates