Learn More
Merck Millicell-ERS2 Volt-Ohm Meter and accessories
Product Code. 11740363 Shop All Merck Products
Click to view available options
Product Type:
Adjustable Electrode
Fixed Test Electrode
Millicell Ers 2V Ohm Meter
Specialized Electrode
Unit Size:
Each
Product Code. 11740363
Brand: Merck MERSSTX00
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Includes
Millicell-ERS2 comes with meter, battery, STX01 electrode, 1,000 Ω test electrode, battery charger. Specialized electrodes for Millicell-96 well plate are available (Cat. No. MERSSTX00).
Measures Trans Epithelial Electrical Resistance (TEER) of epithelial cells in culture
Due to the small size of the electrodes, the user can easily measure transepithelial voltage and the resistance of cells grown on microporous membranes.
Services
Electrodes and batteries are warranted for 30 days only from the date on which the customer receives these items
TRUSTED_SUSTAINABILITY
Specifications
Product Type Specialized Electrode
Length (English) 7.3 in.
Length (Metric) 19 cm
For Use With (Application) Millicell-96 well plate specialized electrodes for volt-ohmmeter readings
Height (English) 2.3 in.
Height (Metric) 6 cm
Hertz 12.5 Hz
Humidity 0 to 90%
Includes Specialized electrodes for millicell-96 well plate
No. per Pack 1 Pair
Run Time 8 to 10 hr.
Temperature Range (English) 50° to 100°F
Temperature Range (Metric) 10° to 38°C
Weight (English) 3 lb.
Weight (Metric) 1.4 kg
Width (Metric) 11 cm