Learn More
Merck MultiScreen™ Carrier Racks
Shop All Merck Products
Click to view available options
Product Type:
Carrier Rack for 12 x 75mm Tubes
Carrier Rack for 4mL Vials
Carrier Rack for 7mL Vials
Unit Size:
Each
Description
- Carrier racks hold sample collection containers in place; available for 4mL vials, 7mL vials, or 12 x 75mm tubes (12 racks per carrier rack assembly)
Specifications
Specifications
| Product Type | Carrier Rack for 4mL Vials |
| For Use With (Application) | Radionucleotide counting by transfer of individual filters to tubes |
| Quantity | 1/Pk. |