Learn More
Merck Replacement Solutions for Montage™ SEQ96 and SEQ384 Sequencing Reaction Cleanup Kits
Click to view available options
Product Type:
Injection Solution
Wash Solution
Unit Size:
500mL
Description
Montage SEQ kits provide efficient, centrifugation-free sequencing reaction cleanup in 96 and 384-well formats. The unique size-exclusion separation technology eliminates the need for costly gel filtration resins or bind-wash-elute columns.
Specifications
Specifications
| For Use With (Equipment) | Montage™ SEQ96 and SEQ384 sequencing reaction cleanup kits |
| For Use With (Application) | Plasmid Miniprep, BAC Prep |
| Product Type | Wash Solution |
| Quantity | 500 mL |