Learn More
Merck SNAP i.d.™ 2.0 Protein Detection System: Mini and Midi Blot Holding Frames
Shop All Merck Products
Click to view available options
Type:
Midi Blot Holding Frames
Mini Blot Holding Frames
Quantity:
1/Pk.
2/Pk.
Unit Size:
Each
Pack of 2
Description
For use with EMD Millipore SNAP i.d. 2.0 Protein Detection System (Cat. No. SNAP2MM)
Specifications
Specifications
| For Use With (Equipment) | Snap i.d.™ 2.0 Protein Detection System-mini |
| Type | Mini Blot Holding Frames |
| Includes | Clear lids, easy locking latch, umbrella valve |
| Quantity | 1/Pk. |
| Stackable | Yes |