missing translation for 'onlineSavingsMsg'
Learn More

Syngene™ Emission Filter

Syngene™ Emission Filter is for use in Syngene Gbox and Gbox Mini Systems 7 position filter wheel

Brand:  Syngene™ FILTLY800-GXM

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 14804811

  • 1890.00€ / Each
Estimated Shipment: 03-06-2024
to see stock.
Add to basket

Description

Description

  • For use with Gbox and Gbox Mini Systems 7 position filter wheel
Specifications

Specifications

1
800 nm
Filter
Gbox and Gbox Mini
7 Position Filter Wheel, Multiplex Suitable
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions