missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Emtricitabine L-Menthyl Ester ((2R,5S)-5-(4-Amino-5-fluoro-2-oxo-1(2H)-pyrimidinyl)-1,3-oxathiolane-2-carboxylic Acid (1R,2S,5R)-5-Methyl-2-(1-methylethyl)cyclohexyl Ester), Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
Description
Emtricitabine L-Menthyl Ester ((2R,5S)-5-(4-Amino-5-fluoro-2-oxo-1(2H)-pyrimidinyl)-1,3-oxathiolane-2-carboxylic Acid (1R,2S,5R)-5-Methyl-2-(1-methylethyl)cyclohexyl Ester)
Specifications
Specifications
| Chemical Name or Material | Emtricitabine L-Menthyl Ester |
| CAS | 764659-72-5 |
| Analyte or Component Names | Emtricitabine L-Menthyl Ester |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H26 F N3 O4 S |
| Certifications/Compliance | ISO 17025 |
| Synonym | (2R,5S)-5-(4-Amino-5-fluoro-2-oxo-1(2H)-pyrimidinyl)-1,3-oxathiolane-2-carboxylic Acid (1R,2S,5R)-5-Methyl-2-(1-methylethyl)cyclohexyl Ester, Emtricitabine L-Menthyl Ester |
| InChI Formula | InChI=1S/C18H26FN3O4S/c1-9(2)11-5-4-10(3)6-13(11)25-16(23)17-26-14(8-27-17)22-7-12(19)15(20)21-18(22)24/h7,9-11,13-14,17H,4-6,8H2,1-3H3,(H2,20,21,24)/t10-,11+,13-,14+,17-/m1/s1 |
| SMILES | CC(C)[C@@H]1CC[C@@H](C)C[C@H]1OC(=O)[C@@H]2O[C@@H](CS2)N3C=C(F)C(=NC3=O)N |
| IUPAC Name | [(1R,2S,5R)-5-methyl-2-propan-2-ylcyclohexyl] (2R,5S)-5-(4-amino-5-fluoro-2-oxopyrimidin-1-yl)-1,3-oxathiolane-2-carboxylate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction