missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Viral 136R/Y136 Protein

Product Code. 18768587
Click to view available options
:
25μg
This item is not returnable. View return policy

Product Code. 18768587

Brand: R&D Systems™ 8976BR025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Viral 136R/Y136 Protein is derived from CHO. The Recombinant Viral 136R/Y136 Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Bioactivity
Molecular Weight (g/mol) Observed M.W.: 88-120kDa, reducing conditions, Theoretical M.W.: 41kDa
Quantity 25μg
Source Chinese Hamster Ovary cell line,CHO-derived viral 136R protein, (N-terminus) Viral 136R/Y136 (Tyr17-Lys351) Accession &Num; Q9DHH7 HPGGGSGGGSGGGS HHHHHH (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Common Name 136R
Biological Activity Measured by its ability to inhibit Type-I and Type-III IFN-mediated anti-viral activity. Huang, J. et al. (2007) PANS 104:9822. The ED50 for this effect is typically 2-12ng/mL in the presence of 20 pg/mL Recombinant Human IFN-beta.
Conjugate Unconjugated
Species Virus
Recombinant Recombinant
Expression System Chinese Hamster Ovary cell line, CHO-derived Viral 136R/Y136 (Tyr17-Lys351) Accession # Q9DHH7, N-terminus; HPGGGSGGGSGGGS; HHHHHH, C-terminus
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.