missing translation for 'onlineSavingsMsg'
Learn More
Learn More
endo-N-Hydroxy-5-norbornene-2,3-dicarboximide, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Descripción
It decreases racemization in peptide synthesis and inhibits formation of N-acylureas.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It decreases racemization in peptide synthesis and inhibits formation of N-acylureas.
Solubility
Soluble in ethanol
Notes
Store at room temperature. Incompatible with oxidizing agents.
It decreases racemization in peptide synthesis and inhibits formation of N-acylureas.
Solubility
Soluble in ethanol
Notes
Store at room temperature. Incompatible with oxidizing agents.
Especificaciones
Melting Point | 166°C to 170°C |
Quantity | 10 g |
Beilstein | 13540 |
Solubility Information | Soluble in ethanol |
Formula Weight | 179.18 |
Percent Purity | 97% |
Chemical Name or Material | endo-N-Hydroxy-5-norbornene-2,3-dicarboximide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'