missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Endothall

Product Code. 15443629
Click to view available options
Quantity:
20 mg
100 mg
Unit Size:
100mg
20mg
This item is not returnable. View return policy

Product Code. 15443629

Brand: Thermo Scientific Chemicals J60948.F+

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

A moderately potent inhibitor of protein phosphatase 2A

Endothall is used as an inhibitor of PP2A phosphatase activity. It is also used as a defoliant. It is used as a cell adhesion inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 145-73-3
Molecular Formula C8H10O5, C8H8Na2O5
Synonym 7-Oxabicyclo[2.2.1]heptane-2,3-dicarboxylic acid

Specifications

Quantity 20 mg
Description White
Content And Storage 2°C to 8°C
For Use With (Application) A moderately potent inhibitor of protein phosphatase 2A
Formula Weight 186.20g/mol
Product Type Signal Transduction Reagent
Form Powder
InChI Key XRHVZWWRFMCBAZ-UHFFFAOYNA-L
SMILES [Na+].[Na+].[O-]C(=O)C1C2CCC(O2)C1C([O-])=O
IUPAC Name disodium 7-oxabicyclo[2.2.1]heptane-2,3-dicarboxylate
Molecular Weight (g/mol) 230.13
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.