Learn More
DiscoverX Endothelin-1
Product Code. 17332489
Encompass
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Product Code. 17332489
Brand: DiscoverX 921164
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Endothelin-1 is an agonist for EDNRA and EDNRB. Provided as 10μg of dry powder alongside reconstitution buffer.
Endothelin 1 is an endothelium derived vasoconstrictive peptide produced in vascular endothelial cells. It is one of the three isoforms of endothelin, and a potent agonist of the endothelin receptors type A and type B (EDNRA/ETA and EDNRB/ETB). Widely expressed in all tissues, and responsible for maintenance of vascular tone. Inhibitors and antagonists are used to treat pulmonary hypertension. Provided as 10μg of dry powder alongside reconstitution buffer.
Specifications
Product Type Control Ligands
Quantity 10 μg