missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Endoxifen

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Endoxifen is an estrogen receptor α (ERα) ligand; potent antiestrogen. Metabolite of tamoxifen (Cat. No. 0999). Primary metabolite responsible for the effectiveness of tamoxifen in ER-positive breast cancer.
Specifications
Specifications
| Purity | >98% |
| CAS | 110025-28-0 |
| Quantity | 50 mg |
| Product Type | Endoxifen |
| Molecular Weight (g/mol) | 373.49 |
| Inhibitors | Endoxifen |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction