missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Enoximone
GREENER_CHOICE

Product Code. 18045122
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18045122

Brand: Tocris Bioscience™ 1693/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

PDE3 inhibitor

Inhibitor of type III phosphodiesterase (PDE3). Increases intracellular cyclic AMP (cAMP) concentrations and enhances myocardial contractility. Also induces concentration-dependent vasodilation in vitro.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77671-31-9
Molecular Formula C12H12N2O2S

Specifications

Quantity 10 mg
Formula Weight 248.3
Purity >99%
Molecular Weight (g/mol) 248.30
Chemical Name or Material Enoximone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.