Learn More
Memmert™ Entry Port 100 mm for UN260
Product Code. 17589985 Shop All Memmert Products
Change view
Click to view available options
Diameter (Metric):
100 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
17589985 100 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17589985 Supplier Memmert™ Supplier No. UN260F9
Only null left
Add to basket
Add to basket
Optional entry port for Memmert Ovens.
  • Entry port with 100 mm diameter
  • Flap closure
  • Custom position TBC in the back
Specifications
For Use With (Equipment) UN260 Oven
Type Entry Port
Diameter (Metric) 100 mm