Learn More
Memmert™ Entry Port 14 mm for IN260
Product Code. 17547545 Shop All Memmert Products
Change view
Click to view available options
Diameter (Metric):
14 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
17547545 14 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17547545 Supplier Memmert™ Supplier No. IN260D6
Only null left
Add to basket
Add to basket
Optional entry port for Memmert Incubators.
  • Entry port with 14 mm diameter
  • Flap closure
  • Custom position TBC in the back
Specifications
For Use With (Equipment) IN260 Incubator
Type Entry Port
Diameter (Metric) 14 mm