missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Epidermal Growth Factor Receptor/EGFR Ab-1, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in affinity purification, immunoprecipitation and flow cytometry experiments.

139.00€ - 657.90€

Specifications

Antigen Epidermal Growth Factor Receptor /EGFR Ab-1
Clone 528
Applications Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Conjugate Concentration Quantity Price Quantity & Availability  
Product Code Brand Conjugate Concentration Quantity Price Quantity & Availability  
12660747
View Documents Promotion Details
Epredia™
MS-268-P0
Unconjugated 200μg/mL 100 μL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
12690747
View Documents Promotion Details
Epredia™
MS-268-PABX
Unconjugated 1mg/mL 200 μL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
Description

Description

EGFR is a type I receptor tyrosine kinase with sequence homology to erbB-1, -2, -3 -4 or HER-1, -2, -3 -4. It binds to Epidermal Growth Factor (EGF), Transforming Growth Factor-a (TGF-a), Heparin-binding EGF (HB-EGF), amphiregulin, βcellulin and epiregulin. EGFR is overexpressed in tumors of breast, brain, bladder, lung, gastric, head & neck, esophagus, cervix, vulva, ovary, and endometrium. It is predominantly present in squamous cell carcinomas.

Host Species: Mouse

Clone: 528

Isotype: IgG2a

Species Reactivity: Human and Dog. Does not react with mouse and rat. Others not known.

Epitope: Extracellular domain

Immunogen: Purified EGFR from A431 cells

Molecular Weight: 170 kDa (wild type) and 145 kDa (vIII variant)

Positive Control: A431 cells

Cellular Localization: Membrane

Recommended for:

  • Western Blotting (Not suitable)
  • Affinity Purification
  • Blocks EGF/TGFα-induced Activation of EGFR
  • Inhibits Binding of EGFR Ab-2 (clone 225) to EGFR
  • Inhibits Proliferation of A431 Cells
  • Arrests Tumor Growth in vivo
  • Immunoprecipitation (Native only)
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Epidermal Growth Factor Receptor /EGFR Ab-1
Flow Cytometry, Immunoprecipitation
Unconjugated
Mouse
EGFR Family
Purified EGFR from A431 cells
Primary
528
Monoclonal
Concentrated
RUO
Canine, Human
IgG2a
Videos
SDS
Documents

Documents

Product Certifications