Learn More
Tocris Bioscience™ Eplivanserin hemifumarate
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Eplivanserin hemifumarate is a potent and selective 5-HT2A antagonist (IC50 values are 5.8 and 120 and >100 nM for 5-HT2A, 5-HT2B and 5-HT2C, respectively). Attenuates cocaine-induced hyperactivity. Increases dopamine (DA) release in rat medial prefrontal cortex (mPFC); potentiates haloperidol-induced DA release in the mPFC and nucleus accumbens.
Specifications
Specifications
| Chemical Name or Material | (1Z,2E)-1-(2-Fluorophenyl)-3-(4-hydroxyphenyl)-2-propen-1-one O-[2-(dimethylamino)ethyl]oxime hemifumarate |
| CAS | 130580-02-8 |
| Synonym | SR-46349 hemifumarate, SR-46349B |
| Quantity | 50 mg |
| Target | 5-HT2A Receptor Antagonists |
| Molecular Formula | C19H21FN2O2.0.5C4H4O4 |
| Formula Weight | 386.42 |
| Purity | >98% |