Learn More
STEMCELL Technologies EPO Immunoaffinity Isolation Plate
Product Code. 17147711
Encompass_Preferred
Click to view available options
Quantity:
1 Plate
Unit Size:
1 set
Product Code. 17147711
Brand: STEMCELL Technologies 01640
Antibody-coated microtiter plate for immunoaffinity isolation of human EPO
  • EPO Immunoaffinity Isolation Plate is a 96-well microtiter plate coated with a high-affinity monoclonal antibody (anti-EPO-26) against human erythropoietin (EPO)
  • Each well can be used for immunoaffinity purification of natural or recombinant human EPO from serum, plasma, urine or other samples
  • EPO isolated using the EPO Immunaffinity Isolation Plate is appropriate for further analysis by methods such as SDS-PAGE, isoelectric focusing and mass spectrometry
Specifications
For Use With (Application) ELISA
Quantity 1 Plate