Learn More
STEMCELL Technologies EPO Immunoaffinity Isolation Plate
Click to view available options
Quantity:
1 Plate
Unit Size:
1 set
Description
- EPO Immunoaffinity Isolation Plate is a 96-well microtiter plate coated with a high-affinity monoclonal antibody (anti-EPO-26) against human erythropoietin (EPO)
- Each well can be used for immunoaffinity purification of natural or recombinant human EPO from serum, plasma, urine or other samples
- EPO isolated using the EPO Immunaffinity Isolation Plate is appropriate for further analysis by methods such as SDS-PAGE, isoelectric focusing and mass spectrometry
Specifications
Specifications
| For Use With (Application) | ELISA |
| Quantity | 1 Plate |