Learn More
Invitrogen™ EPO Receptor Human ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ EHEPORX10
Additional Details : Weight : 2.50000kg
Description
The Human EPO Receptor (EPO R) ELISA quantitates Hu EPO R in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu EPO R. Principle of the method The Human EPO R solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Recombinant Human Erythropoietin Receptor (EPO R) is a type I membrane-spanning protein, expressed in a mouse myeloma cell line, NSO. This product is produced from a cDNA sequence encoding the extracellular domain of the EPO receptor. The 225 amino acid residue mature recombinant soluble EPO receptor, generated after cleavage of a 25 amino acid residue signal peptide, has predicted molecular mass of approximately 25 kDa.Specifications
P19235 | |
180 pg/mL | |
Solid-phase sandwich ELISA | |
ELISA Kit | |
Plasma, Serum, Supernatant | |
10 x 96 assays | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
2057 | |
<12% | |
Biotinylated Detection Antibody, Adhesive Plate Covers, Standard, Chromogen, Stop Solution, Pre-coated 96 well plate, Wash Buffer, SAV-HRP, Assay Diluent concentrate | |
10 x 96 Tests | |
Signal Transduction | |
2°C to 8°C | |
Human | |
4 hr. 45 min. |
187.5 to 12,000 pg/mL | |
180 pg/mL to 12,000 pg/mL | |
Biotin | |
Other Proteins | |
This ELISA kit shows no cross-reactivity with the following cytokines tested: human Angiogenin, BDNF, BLC, CNTF, ENA-78, FGF-4, IL-1 A, IL-1 b, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IL-11, IL-12 p70, IL-12 p40, IL-13, IL-15, IL-309, IP-10, FGF-4, FGF-6, FGF-7, G-CSF, GDNF, GM-CSF, IFN-g, IGFBP-2, IGFBP-3, IGFBP-4, Leptin (OB), MCP-1, MCP-2, MCP-3, MDC, MIF, MIG, MIP-1 A, MIP-1 b, MIP-1 delta, PARC, PDGF, RANTES, SCF, SDF-1 alpha, TARC, TGF-b, TIMP-1, TIMP-2, TNF-a, TNF-b, TPO, VEGF. | |
Human | |
Colorimetric Microplate Reader | |
EPOR | |
<10% | |
HRP | |
RUO | |
Plasma, 50 μL; Serum, 50 μL; Supernatant, 100 μL | |
EPO-R | |
1 hr. 20 min. |