Learn More
Eppendorf™ S-4-104 Rotor Adapters
Shop All Eppendorf Products
Click to view available options
Capacity:
1 x 250 mL Tubes
1 x 500 mL Bottle
14 x 15 mL Conical Tubes
14 x 5 mL Conical Tubes
20 x 5.5/12 mL Tubes
23 x 2.6/8 mL Tubes
23 x 7/17 mL Tubes
4 x 750 mL Wide-mouth Bottles
5 x 50 mL Conical Tubes
50 x 1.5/2 mL Tubes
7 x 50 mL Conical Tubes
Unit Size:
1 set
Pack of 2
Description
Includes
2 piece set
- For use with Centrifuge 5810/5810 R
Specifications
Specifications
| Type | Rotor Adapter |
| Capacity | 23 x 2.6/8 mL Tubes |
| For Use With (Equipment) | S-4-104 rotor for 5810/5810R centrifuge |
| No. per Pack | 2 Pack |