missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ ThermoMixer™ C Accessory: Smartblock™ Thermoblocks

Maintain accurate uniform temperatures with these thermoblocks. Eppendorf™ Smartblock™ Thermoblocks are compatible with Eppendorf ThermoMixer™ C, and support a broad range of applications.

Brand:  Eppendorf™ 5367000025

Additional Details : Weight : 0.58500kg

 View more versions of this product

Product Code. 13578470

  • 543.00€ / Each


Estimated Shipment
10-06-2020
Add to basket

Description

Description

 
  • Exchangeable in just seconds
  • Automatic block recognition for safe handling

 

Specifications

Specifications

Accessory, Block
Eppendorf ThermoMixer C
1.5 to 2mL x 24 Cryotubes
12.5 mm
24 x 1.5 to 2.0mL
Videos
Documents