missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ twin.tec™ 384-Well Microbiology PCR Plates

Continuous skirt for compatibility with automated systems

138.00€

Specifications

Format Standard
Array 16 x 24
For Use With (Equipment) Automated systems
Skirt Style Skirted
Material Polypropylene
View More Specs

Products 2
Product Code Brand Color Price Quantity & Availability  
Product Code Brand Color Price Quantity & Availability  
10023927
View Documents Promotion Details
Eppendorf™
0030129342
Clear
138.00€
Pack of 10
View alternative products
Save Up to 
»
Estimated Shipment: 29-04-2024
to see stock.
Add to basket
 
10267467
View Documents Promotion Details
Eppendorf™
0030129350
Blue
138.00€
Pack of 10
View alternative products
Save Up to 
»
Estimated Shipment: 29-04-2024
to see stock.
Add to basket
 
Description

Description

  • Skirted for bar code
  • Stackable
  • Within SBS footprint recommendations
  • Ideal for most common 384-well thermocyclers, particularly for Eppendorf Mastercycler™ Pro 384
  • 45 to 40μL max. well volume

Specifications

Specifications

Standard
Automated systems
Polypropylene
384
10/Pk.
16 x 24
Skirted
40 to 45 μL
Round Bottom (U-well)
Videos
SDS
Documents

Documents

Product Certifications